Anti-NFKB p65(acetyl-K310) Rabbit Polyclonal Antibody
Catalog # 77439-884
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:NFKB p65(acetyl-K310)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Gene ID:5970
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Storage temperature:Store at −20 °C for 12 months
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Cat. no.:77439-884
- Packaging:Vial
Specifications
About this item
NF-kappa-B is a ubiquitous transcription factor involved in several biological processes. It is held in the cytoplasm in an inactive state by specific inhibitors. Upon degradation of the inhibitor, NF-kappa-B moves to the nucleus and activates transcription of specific genes. NF-kappa-B is composed of NFKB1 or NFKB2 bound to either REL, RELA, or RELB. The most abundant form of NF-kappa-B is NFKB1 complexed with the product of this gene, RELA. Four transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
Type: Primary
Antigen: NFKB p65(acetyl-K310)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse