ContactUsLinkComponent
VWR® Traceable® Four-Channel Alarm Timer with Clock
VWR® Traceable® Four-Channel Alarm Timer with Clock
Catalog # 62344-641
Supplier:  VWR International
undefined
VWR® Traceable® Four-Channel Alarm Timer with Clock
Catalog # 62344-641
Supplier:  VWR International
Specifications
  • Cat. no.:
    62344-641
  • W×H×D:
    7×6.4×1.3 cm (2¾×2½×½")
Specifications
About this item
Microcomputer chip permits setting four separate channels for simultaneous countup or countdown procedures.
  • Massive ³/₄" high display allows viewing from across the room
  • Extra-loud, high decibel alarms for 1 minute and may be silenced manually
  • Timing capacity to 99 hours, 59 minutes and 59 seconds with a 1 second resolution
  • Remarkable memory returns display to previously programmed countdown time at the touch of a button
Large 1.9 cm LCD permits viewing from across the lab. Additional features include stopwatch, time-out, and time-of-day display. Use all four channels simultaneously as count-up stopwatches or in any combination of countup/countdown. The touch of a button returns the display to the previously programmed countdown time. Extra-loud, high-decibel alarm sounds for one minute or may be silenced manually. Finger-size keys make it easy to set times and change channels. Magnetic back allows timer to be placed on any metal surface. Unit also has a flip-open easel for standing on the lab bench, a spring fastener for clipping to a notebook/clipboard, and a hole for use with a lanyard.
When zero is reached the timer starts counting up to show time elapsed since alarming.
Ordering information: Supplied with a one-year, 1.5 V silver-oxide battery. Replacement battery is available separately.
Delivery information: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.