VWR® Timer Plus
5.1×3.8×1.9 cm (2×1½×¾"), 14.2 g (0.5 oz.)
Catalog # 62379-604
Supplier:   VWR International
VWR® Timer Plus
Catalog # 62379-604
Supplier:  VWR International
VWR® Timer Plus
VWR® Timer Plus
5.1×3.8×1.9 cm (2×1½×¾"), 14.2 g (0.5 oz.)
Price
$122.82
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Key chain timer is three timers in one - countdown alarm timer and stopwatch.
  • Traceable®
  • Six-digit LCD display
  • Programmable timer functions
  • VWR® six-year limited warranty
Ideal for catching test results and signaling time intervals and appointments.
Six-digit LCD always shows time in hours, minutes, and seconds. Countdown mode can be programmed in one-minute increments from 100 hours to 1 minute. When display reaches zero, a loud, beeping alarm sounds for 60 seconds. The alarm can be silenced manually. In stopwatch mode, it times up from 1 second to 99 hours and then repeats. The timer has a time-out feature in both countup and stopwatch modes.
Clip it to a button-hole, belt loop, or notebook.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with an eighteen-month continuous-use replaceable silver-oxide battery, keychain, lanyard, and magnet for placing on metal.
Spécifications
  • W×H×D:
    5.1×3.8×1.9 cm (2×1½×¾")
  • Weight:
    14.2 g (0.5 oz.)