Innovating Science Unknown Compositions Samples: Ferrous Ammonium Sulfate
Catalog # 470354-898
Supplier: Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Unknown set, ferrous ammonium sulfate
- Size:100 g (sample)
- Cat. no.:470354-898
- No. of Uses:20 samples
Specifications
About this item
A series of ferrous ammonium sulfate samples with a range of known purities.
- 20×100 g samples with purity concentrations ranging from 0.1 to 13% iron
- Reference sheet included
Use these in practicing analytical techniques such as gravimetric, potentiometric, and spectrophotometric analyses.