VWR® 60 Memory, Multi-Function Stopwatch with Countdown
6.7×5.4 cm (2⁵/₈×2¹/₈"), 0.001%
Catalog # 62379-200
Supplier:   VWR International
VWR® 60 Memory, Multi-Function Stopwatch with Countdown
Catalog # 62379-200
Supplier:  VWR International
VWR® 60 Memory, Multi-Function Stopwatch with Countdown
VWR® 60 Memory, Multi-Function Stopwatch with Countdown
6.7×5.4 cm (2⁵/₈×2¹/₈"), 0.001%
Price
$184.58
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Provides repeat functions of countdown alarm timing from 10 hours to 1 second.
  • Traceable®
  • 60 memory settings
  • Programmable
When zero is reached, the timer alarms and begins to count down again from the programmed time.
Three-line display simultaneously shows running time, cumulative split, and interval split. Sixty memories recall all splits, display sample numbers 1 through 60, and show longest, shortest, and average interval splits. Stopwatch times to 9 hours, 59 minutes, 59 seconds, 99 hundredths.
Repeat cycle continues automatically until stopped. Display continually shows running time remaining to zero and cycles counted from 1 to 60.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with lanyard, instructions, and two-year, 3 V lithium battery.
Spécifications
  • Dimensions:
    6.7×5.4 cm (2⁵/₈×2¹/₈")
  • Accuracy:
    0.001%