VWR® Three-Button Stopwatch
5.4 cm (2¹/₈"), 0.1%
  62379-234
VWR® Three-Button Stopwatch
  62379-234
VWR® Three-Button Stopwatch
VWR® Three-Button Stopwatch
5.4 cm (2¹/₈"), 0.1%
$62.49
Each
For easy-duty, occasional use where high precision is not a requirement.
  • Traceable®
  • Designed for occasional, easy-duty use
  • Multiple modes of functionality
  • Audible confirmation of button pressing
Unit times to 24 hours, with a resolution of 0.01 second for thirty minutes and one second thereafter. Modes include start/stop, time-out, cumulative split, and continuous roll-over timing. Also displays time and date. Provides a sound confirmation each time a button is pressed.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with lanyard and 1.5 V silver-oxide battery.
  • Ø:
    5.4 cm (2¹/₈")
  • Accuracy:
    0.1%