SP Bel-Art Dispensing/Drop Bottles, LDPE, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Low-density polyethylene bottles and spouts have sealer caps that snap on easily for a tight seal.
- LDPE construction
- Tight seal
- Caps snap on for easy closure
Spout openings are standard 0.76 mm (0.03"). Opening can be enlarged by cutting back with scissors.
Frequently Bought Together