ContactUsLinkComponent
PYREX® VISTA™ Standard Low Griffin Beakers, Corning
  89090-436
 :  Corning
 : 70000-30
Product available on GSA Advantage®
Low Manufacturing Impact
undefined
PYREX® VISTA™ Standard Low Griffin Beakers, Corning
  89090-436
 :  Corning
 :  70000-100
 : 70000-30
Product available on GSA Advantage®
  Low Manufacturing Impact
Select an Option
5 - 25 ml
Select an Option
30 ml (1 oz.)
Select an Option
89090-436
Select an Option
70000-30
  • Catalog No:
  • 89090-436
  • 89090-434
  • 89090-432
  • 89090-430
  • 89090-428
  • 89090-426
  • 89090-424
  • 89090-442
  • 89090-440
  • 89090-438
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
These beakers with spout are made of Type I Class A low expansion borosilicate glass, per ASTM E438.
  • High mechanical strength
  • Resistance to thermal shock
Beakers have blue graduations and an extra large blue marking spot. The 250 through 1000 ml beakers have a double graduated metric scale to indicate approximate content.
*10 ml beaker is not graduated
 : ASTM E-960
Frequently Bought Together