ContactUsLinkComponent
VITLAB® Griffin Beakers, Polypropylene, BrandTech
Catalogue # 83008-826
Fournisseur:  Brandtech
Fournisseur #: 616081
undefined
VITLAB® Griffin Beakers, Polypropylene, BrandTech
Catalogue # 83008-826
Fournisseur:  Brandtech
Supplier Number:  605081
Fournisseur #: 616081
S'identifier pour la disponibilité
Select an Option
170 mm
Select an Option
250 ml
Select an Option
3 L (101.4 oz.)
Select an Option
212 mm (8.35")
Select an Option
83008-826
Select an Option
V616081
À propos de cet article
High quality beakers feature molded graduations (except 83008-814) with blue-screened overprint for legibility.
  • Highly chemically resistant polypropylene
  • High translucency
  • Blue screened graduations provide high contrast with many liquids
Beakers are translucent for compatibility and offer good chemical and break resistance.
Certifications: All beakers meet the requirements of ISO 7056.
Frequently Bought Together