ContactUsLinkComponent
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Catalog # 83008-742
Supplier:  Brandtech
Supplier #: 442081
undefined
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Catalog # 83008-742
Supplier:  Brandtech
Supplier Number:  445081
Supplier #: 442081
Log in for availability
Select an Option
20 ml
Select an Option
1 L (34 oz.)
Select an Option
83008-742
Select an Option
V442081
About this item
Made of durable, translucent polypropylene.
  • Pitchers with molded graduations areavailable with or without overprint
  • Nesting pitchers have screened overprint
  • Blue printing provides high contrast with many liquids
Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.
Frequently Bought Together