ContactUsLinkComponent
Nalgene® Graduated Beakers with Handle, High-Density Polyethylene, Thermo Scientific
Catalogue # 13916-955
Fournisseur #: 1220-1250
undefined
Nalgene® Graduated Beakers with Handle, High-Density Polyethylene, Thermo Scientific
Catalogue # 13916-955
Supplier Number:  1220-1250
Fournisseur #: 1220-1250
S'identifier pour la disponibilité
Select an Option
100 - 1000 ml
Select an Option
1 L (34 oz.)
Select an Option
13916-955
Select an Option
1220-1250
À propos de cet article
Made from high-density polyethylene with extra heavy wall
  • Graduated in both ounces and milliliters
  • Large rectangular handle
Especially useful in handling acids, alkalies, and photographic chemicals.
Frequently Bought Together