Fold and Snap Tube Rack, 13 mm, 90-Place
Fournisseur: VWR International
Certificats
À propos de cet article
Economical durable rack.
- Polypropylene racks ship flat in one piece
- Easy-to-assemble, just fold and snap together
- Stable design can be used in water baths
- Open design allows samples to be incubated at the same temperature
- Color code for easy identification
- Easy to assemble just fold and snap together
The Fold and snap test tube rack is economical and durable. It is easy to assemble, simply fold and snap together. The rack is made out of polypropylene.
Frequently Bought Together