Fold and Snap Tube Racks, 17 mm, 60-Place
Economical durable rack.
- Polypropylene racks ship flat in one piece
- Easy-to-assemble, just fold and snap together
- Stable design can be used in water baths
- Open design allows samples to be incubated at the same temperature
- Color code for easy identification
- Easy to assemble just fold and snap together
The Fold and Snap Test tube rack is economical and durable.
Frequently Bought Together