ContactUsLinkComponent
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
  77395-378
undefined
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
  77395-378
 :  HS23224
  • Color:
    Blue
  • Material:
    HDPE/carbon steel
  • Cat. no.:
    77395-378
  • For Vials:
    Tube array: 2×3; wells: 6; well Ø: 55 - 60 mm
Chemically resistant HDPE coated wire racks.
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
The HDPE coated wire rack is durable for transport or sample handling. It is also chemically resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.