Tube Rack, Polystyrene Friction - Fit
Catalogue # 77395-372
Fournisseur: Heathrow Scientific
Spécifications
- Color:Blue
- Material:PS
- Cat. no.:77395-372
- For Tubes:50 ml tubes, 1 Well (29 mm well Ø)
Spécifications
À propos de cet article
One-well friction-fit tube rack for 50 ml tubes.
- Made from polystyrene
- Open design enables easy identification of samples
- Holds conical and round bottom tubes
- Slides smoothly across the benchtop
One-well wire tube rack holds tubes for small space requirements. The rack accommodates conical and round-bottom tubes.