Microscope Slide Boxes, 25-Place Foam
Supplier: VWR International
Jump to:
À propos de cet article
Easily access slides with friction-fit cover.
- Organize and identify; durable and safe
- Lining and slots are numbered to correspond to the slide inventory sheet
- Organize by 'Project' and 'Date' with pre-molded labeling area
- Available in multiple colors for project coding
- ABS plastic boxes available with either cork or foam lining
- Designed to securely store slides in grooved slots
- Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides (25×75 mm and 1×3").
3 Options Available Below
Product Details & Documents
Easily access slides with friction-fit cover.
- Organize and identify; durable and safe
- Lining and slots are numbered to correspond to the slide inventory sheet
- Organize by 'Project' and 'Date' with pre-molded labeling area
- Available in multiple colors for project coding
- ABS plastic boxes available with either cork or foam lining
- Designed to securely store slides in grooved slots
- Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides (25×75 mm and 1×3").
Product Family Options
Informations sur le produit
- MatièreCapacitéColorAvailabilityPrice
- ABS25 - Place (26× 76 mm and 1× 3 in slide size)BlueLog in pour la disponibilitéCase of 72Case of 72 - $1,395.65$1,395.65Prix promotionnel pour les nouveaux clientsEach$19.38Case of 72$1,395.65
- ABS25 - Place (26× 76 mm and 1× 3 in slide size)RedLog in pour la disponibilitéCase of 72Case of 72 - $1,395.65$1,395.65Prix promotionnel pour les nouveaux clientsEach$19.38Case of 72$1,395.65
- ABS25 - Place (26× 76 mm and 1× 3 in slide size)WhiteLog in pour la disponibilitéCase of 72Case of 72 - $1,395.65$1,395.65Prix promotionnel pour les nouveaux clientsEach$19.38Case of 72$1,395.65