Operating Scissors, Sklar
Supplier:   Sklar
Operating Scissors, Sklar
Supplier:  Sklar
Certificates
Operating Scissors, Sklar
Operating Scissors, Sklar
51 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures, and multiple lengths, these scissors are used for general, multi-purpose cutting, and dissecting
Censitrac Ready OR Grade Operating Scissors are industry standard and constructed of stainless steel.
Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Operating Scissors
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Curvature
    Handle
    Sterility
    Availability
    Price
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $147.13
    $147.13
    Prix promotionnel pour les nouveaux clients
    Each
       $147.13
    Supplier Number: 22-1545
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.56
    $175.56
    Prix promotionnel pour les nouveaux clients
    Each
       $175.56
    Supplier Number: 22-1645
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $125.56
    $125.56
    Prix promotionnel pour les nouveaux clients
    Each
       $125.56
    Supplier Number: 13-1045
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $114.13
    $114.13
    Prix promotionnel pour les nouveaux clients
    Each
       $114.13
    Supplier Number: 13-2045
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.54
    $119.54
    Prix promotionnel pour les nouveaux clients
    Each
       $119.54
    Supplier Number: 14-1045
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $174.31
    $174.31
    Prix promotionnel pour les nouveaux clients
    Each
       $174.31
    Supplier Number: 22-1145
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $116.42
    $116.42
    Prix promotionnel pour les nouveaux clients
    Each
       $116.42
    Supplier Number: 14-2045
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $187.21
    $187.21
    Prix promotionnel pour les nouveaux clients
    Each
       $187.21
    Supplier Number: 22-1245
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $150.41
    $150.41
    Prix promotionnel pour les nouveaux clients
    Each
       $150.41
    Supplier Number: 15-1045
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $198.89
    $198.89
    Prix promotionnel pour les nouveaux clients
    Each
       $198.89
    Supplier Number: 22-1345
  • Operating Scissors, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $116.42
    $116.42
    Prix promotionnel pour les nouveaux clients
    Each
       $116.42
    Supplier Number: 15-2045
  • Operating Scissors, Delicate, Premium OR-Grade
    11.4 cm (4.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.86
    $166.86
    Prix promotionnel pour les nouveaux clients
    Each
       $166.86
    Supplier Number: 22-1445
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.59
    $113.59
    Prix promotionnel pour les nouveaux clients
    Each
       $113.59
    Supplier Number: 13-2050
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $112.40
    $112.40
    Prix promotionnel pour les nouveaux clients
    Each
       $112.40
    Supplier Number: 14-1050
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $117.57
    $117.57
    Prix promotionnel pour les nouveaux clients
    Each
       $117.57
    Supplier Number: 14-2050
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.94
    $115.94
    Prix promotionnel pour les nouveaux clients
    Each
       $115.94
    Supplier Number: 15-1050
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.94
    $115.94
    Prix promotionnel pour les nouveaux clients
    Each
       $115.94
    Supplier Number: 15-2050
  • Operating Scissors, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $117.70
    $117.70
    Prix promotionnel pour les nouveaux clients
    Each
       $117.70
    Supplier Number: 13-1050
  • Operating Scissors, Delicate, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.93
    $170.93
    Prix promotionnel pour les nouveaux clients
    Each
       $170.93
    Supplier Number: 22-1150
  • Operating Scissors, Delicate, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.58
    $171.58
    Prix promotionnel pour les nouveaux clients
    Each
       $171.58
    Supplier Number: 22-1250
  • Operating Scissors, Delicate, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $167.83
    $167.83
    Prix promotionnel pour les nouveaux clients
    Each
       $167.83
    Supplier Number: 22-1350
  • Operating Scissors, Delicate, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $207.28
    $207.28
    Prix promotionnel pour les nouveaux clients
    Each
       $207.28
    Supplier Number: 22-1450
  • Operating Scissors, Delicate, Premium OR-Grade
    12.7 cm (5)"
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $214.78
    $214.78
    Prix promotionnel pour les nouveaux clients
    Each
       $214.78
    Supplier Number: 22-1650
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $217.42
    $217.42
    Prix promotionnel pour les nouveaux clients
    Each
       $217.42
    Supplier Number: 22-1655
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $190.48
    $190.48
    Prix promotionnel pour les nouveaux clients
    Each
       $190.48
    Supplier Number: 22-1555
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $120.79
    $120.79
    Prix promotionnel pour les nouveaux clients
    Each
       $120.79
    Supplier Number: 13-1055
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.42
    $115.42
    Prix promotionnel pour les nouveaux clients
    Each
       $115.42
    Supplier Number: 13-2055
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $137.65
    $137.65
    Prix promotionnel pour les nouveaux clients
    Each
       $137.65
    Supplier Number: 14-1055
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $109.74
    $109.74
    Prix promotionnel pour les nouveaux clients
    Each
       $109.74
    Supplier Number: 14-2055
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.05
    $148.05
    Prix promotionnel pour les nouveaux clients
    Each
       $148.05
    Supplier Number: 15-1055
  • Operating Scissors, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $118.35
    $118.35
    Prix promotionnel pour les nouveaux clients
    Each
       $118.35
    Supplier Number: 15-2055
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $177.97
    $177.97
    Prix promotionnel pour les nouveaux clients
    Each
       $177.97
    Supplier Number: 22-1155
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.52
    $170.52
    Prix promotionnel pour les nouveaux clients
    Each
       $170.52
    Supplier Number: 22-1255
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.83
    $210.83
    Prix promotionnel pour les nouveaux clients
    Each
       $210.83
    Supplier Number: 22-1355
  • Operating Scissors, Delicate, Premium OR-Grade
    13.9 cm (5.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.24
    $210.24
    Prix promotionnel pour les nouveaux clients
    Each
       $210.24
    Supplier Number: 22-1455
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $143.27
    $143.27
    Prix promotionnel pour les nouveaux clients
    Each
       $143.27
    Supplier Number: 13-1060
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.59
    $142.59
    Prix promotionnel pour les nouveaux clients
    Each
       $142.59
    Supplier Number: 13-2060
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.56
    $138.56
    Prix promotionnel pour les nouveaux clients
    Each
       $138.56
    Supplier Number: 14-1060
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $141.03
    $141.03
    Prix promotionnel pour les nouveaux clients
    Each
       $141.03
    Supplier Number: 14-2060
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $144.42
    $144.42
    Prix promotionnel pour les nouveaux clients
    Each
       $144.42
    Supplier Number: 15-1060
  • Operating Scissors, Premium OR-Grade
    15.2 cm (6")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $137.45
    $137.45
    Prix promotionnel pour les nouveaux clients
    Each
       $137.45
    Supplier Number: 15-2060
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $112.93
    $112.93
    Prix promotionnel pour les nouveaux clients
    Each
       $112.93
    Supplier Number: 13-1065
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.25
    $115.25
    Prix promotionnel pour les nouveaux clients
    Each
       $115.25
    Supplier Number: 13-2065
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.06
    $142.06
    Prix promotionnel pour les nouveaux clients
    Each
       $142.06
    Supplier Number: 14-1065
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $141.92
    $141.92
    Prix promotionnel pour les nouveaux clients
    Each
       $141.92
    Supplier Number: 14-2065
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $186.06
    $186.06
    Prix promotionnel pour les nouveaux clients
    Each
       $186.06
    Supplier Number: 15-1065
  • Operating Scissors, Premium OR-Grade
    16.5 cm (6.5")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.73
    $138.73
    Prix promotionnel pour les nouveaux clients
    Each
       $138.73
    Supplier Number: 15-2065
  • Operating Scissors, Premium OR-Grade
    18.4 cm (7.25")
    Stainless steel
    Sharp/Sharp, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.81
    $185.81
    Prix promotionnel pour les nouveaux clients
    Each
       $185.81
    Supplier Number: 13-1072
  • Operating Scissors, Premium OR-Grade
    18.4 cm (7.25")
    Stainless steel
    Sharp/Sharp, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $161.49
    $161.49
    Prix promotionnel pour les nouveaux clients
    Each
       $161.49
    Supplier Number: 13-2072
  • Operating Scissors, Premium OR-Grade
    18.4 cm (7.25")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $188.18
    $188.18
    Prix promotionnel pour les nouveaux clients
    Each
       $188.18
    Supplier Number: 14-1072
  • Operating Scissors, Delicate, Premium OR-Grade
    127 mm (5")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $207.28
    $207.28
    Prix promotionnel pour les nouveaux clients
    Each
       $207.28
    Supplier Number: 22-1550