Laboratory Notebook, Black
Laboratory notebook, lined, Black, 100
Catalog # 77395-446
Supplier: Heathrow Scientific
Fournisseur #: HS8610EV
Jump to:
Product Details & Documents
Laboratory notebooks keep information safe and well organized.
- Quality Smyth Sewn binding holds pages securely and lies flat when opened
- Made of acid-free paper, pages are numbered and include space for title, date, project number, book number, date and witness signature
- User data, notebook instructions, table of contents, periodic table, and conversion tables are included in every book
- Durable hardbound, water-resistant cover keeps information safe
- Includes marker ribbon to keep your place
- Foil stamped title for easy identification
The laboratory notebooks are used for a variety of applications such as recording research data, experimental procedures, design, development ideas and more.
Spécifications
- Cover Color:Black
- Désignation:Laboratory notebook, lined
- No. of Pages:100