VITLAB® Forceps, POM, BrandTech
À propos de cet article
Polyoxymethylene (POM) forceps with rounded ends.
- Rounded ends
- Opaque orange-yellow
POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.
Operating temperature: –40 to 130°C (–40 to 266°F)
Caution: Not recommended for use with ester, acids, and oxidizing agents.
4 Options Available Below
Product Details & Documents
Polyoxymethylene (POM) forceps with rounded ends.
- Rounded ends
- Opaque orange-yellow
POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.
Operating temperature: –40 to 130°C (–40 to 266°F)
Caution: Not recommended for use with ester, acids, and oxidizing agents.
Product Family Options
Informations sur le produit
- LongueurAvailabilityPrice
- 145 mm (5¾")Log in pour la disponibilitéLog in pour la disponibilitéPack of 5Pack of 5 - $55.92$55.92Prix promotionnel pour les nouveaux clientsPack of 5$55.92Supplier Number: 68199
- 180 mm (7⅛")Log in pour la disponibilitéLog in pour la disponibilitéPack of 5Pack of 5 - $107.65$107.65Prix promotionnel pour les nouveaux clientsPack of 5$107.65Supplier Number: 68299
- 115 mm (4½")Log in pour la disponibilitéLog in pour la disponibilitéPack of 5Pack of 5 - $51.44$51.44Prix promotionnel pour les nouveaux clientsPack of 5$51.44Supplier Number: 68099
- 250 mm (9⅞")Log in pour la disponibilitéLog in pour la disponibilitéPack of 5Pack of 5 - $186.46$186.46Prix promotionnel pour les nouveaux clientsPack of 5$186.46Supplier Number: 68399