Manometer Brush, Alta Robbins
Nylon, 76.2 cm (30")
17100-145
: COB-30
Durable nylon bristles with an aluminum handle.
Fits tubes with a bore of ±0.63 cm (±1/4").
- Bristle Material:Nylon
- Longueur totale:76.2 cm (30")