Instrument Cleaning Brushes, Integra™ Miltex®
Fournisseur: Miltex
Certificats
À propos de cet article
Brushes are availabe with either nylon or stainless steel bristles to accomodate a wide variety of instrument cleaning tasks.
- Nylon bristles provide effective cleaning of instrument box locks
- Stainless steel bristles are ideal for cleaning rasps, files and burs
Ordering information: Each brush pack contains a set of three brushes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together