Reagent Reservoir, 50 ml, Bulk Wrap, PVC
77395-282
- Sterility:Non sterile
- Material:PVC
- Packed:Bulk wrap
- Cat. no.:77395-282
Reagent Reservoirs by Heathrow Scientific offer a fresh and modern design with enhanced features to streamline pipetting procedures.
- Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
- Graduation markings on the top edge and inner walls allow measurements of liquid to be easily identified and reduce reagent waste
- Pour-off spouts conveniently located on all four corners for easy spill-free pouring and reduced dripping
- Divided reservoir design allows pipetting of two different reagents; maximize recovery if using only single channel and small amount of reagent