ContactUsLinkComponent
MaxQ™ 6000 Digital Incubating and Refrigerating Stackable Orbital Shakers, 240 V
Certificats
undefined
MaxQ™ 6000 Digital Incubating and Refrigerating Stackable Orbital Shakers, 240 V
Spécifications
  • Catalog No:
  • CA89032-316
  • CA89032-320
  • Max. load:
  • 15.9 kg (35 lbs.)
  • 15.9 kg (35 lbs.)
  • Overall dimensions:
  • 69.9W×83.8D×102.9H cm (271/2×33×401/2")
  • 69.9W×83.8D×102.9H cm (271/2×33×401/2")
  • Orbit:
  • Ø 1.9 cm (3/4")
  • Ø 1.9 cm (3/4")