Savant™ SpeedVac™ Modular Vacuum Concentrators, 230 V
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
The Thermo Scientific™ Savant™ SpeedVac™ SPD210 vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aqueous and organic solvents.
- 230 V model
- Kit
- Medium capacity
- Variety of rotors
- Glass safety cover
- PTFE coated aluminum chamber
The SpeedVac™ SPD210 is capable of drying larger quantities of samples at one time and will support your high-volume sample preparation needs.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (76210-564, 76210-568 and 76210-400) are also available, please see separate product page.
Frequently Bought Together