Acid Resistant Refrigerated CentriVap Centrifugal Vacuum Concentrators, 230 V
Fournisseur: Labconco
Certificats
À propos de cet article
The acid-resistant refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature.
- 230 V model
This exclusive concentrator combines a refrigeration system that cools to –4 °C with a heater that warms to 100 °C to achieve the users desired set point temperatures. It also features a corrosion-resistant, Teflon-coated chamber and Hastelloy shaft to protect against samples containing corrosive solvents.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89187-284) is also available, please see separate product page.
Frequently Bought Together