This unique and innovative designed Vortex Mixer is a variable speed vortexing unit with multi-purpose and maxium convenience. Why take time to search for separate mixing heads for various sized tubes when this vortex mixer provides one mixing head that will accomodate tubes ranging from 0.2 mL - 50 mL and a standard assay plate.
- 220 to 240 V model
- Variable speed rotary dial provides pre-set speed control (1000, 2000, 3000 rpm) during vortex mixing
- Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
- Innovative head can be removed for cleaning purposes
- High impact ABS body shell
- US patent
This sleek and modern designed vortex mixer can meet the mixing requirements of the most popular test tubes used in the today’s lab without the need to change mixing head units.
The innovative design makes maximum use of space available providing holding slots for no less than six different sizes of tubes and a standard assay plate, while the center pad area can be used for quick hand-held mixing of a vast range of tubes and vessels.
- Catalog No:
- 76027-578
- 76027-586
- 76027-794
- Speed range:
- 1000 - 3000 rpm
- 1000 - 3000 rpm
- 1000 - 3000 rpm
- Weight:
- 8.6 lbs.
- 8.6 lbs.
- 8.6 lbs.