Copper Strips
Copper strips, 1.25×1.2" pieces
Catalog # 470352-650
Supplier: Avantor
Jump to:
Product Details & Documents
Great for geology hardness tests.
- Pure copper
- Cut to size for mineral and rock ID testing
- 0.1 mm thickness
Specifications
- Désignation:Copper strips
- L×l:1.25×1.2" pieces