ContactUsLinkComponent
Carb-N-Sert® Webster Needle Holder, Integra™ Miltex®, Integra LifeSciences
Carb-N-Sert® Webster Needle Holder, Integra™ Miltex®, Integra LifeSciences
Catalog # 21909-744
Supplier:  Miltex
undefined
Carb-N-Sert® Webster Needle Holder, Integra™ Miltex®, Integra LifeSciences
Catalog # 21909-744
Supplier:  Miltex
Supplier Number:  8-6TC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    127 mm (5")
  • Cat. no.:
    21909-744
  • Supplier No.:
    8-6TC
Specifications
About this item
Needle holder features jaws made of tungsten carbide bonded onto stainless steel to provide precision when using small sutures in micro-surgical and plastic surgery applications
The edges around the box lock feature distinct non-catch bevels to prevent fraying of suture materials. Delicate, smooth jaws with standard pattern. Autoclavable.