ContactUsLinkComponent
Rotators for Cell Culture, 240 V
Fournisseur:  Ace Glass
Certificats
undefined
Rotators for Cell Culture, 240 V
Fournisseur:  Ace Glass
Spécifications
  • Catalog No:
  • 89084-428
  • 89084-430
  • Weight:
  • 17.0 lbs.
  • 17.0 lbs.
  • Load capacity:
  • 5 lbs.
  • 5 lbs.