Micro Surgery Scissors, Integra™ Miltex®
Micro Surgery Scissors, Integra™ Miltex®
15.2 cm (6"), 6 mm
Catalog # 89049-674
Supplier:   Miltex
Supplier #:  17-2150
Micro Surgery Scissors, Integra™ Miltex®
Catalog # 89049-674
Supplier:  Miltex
Micro Surgery Scissors, Integra™ Miltex®
Micro Surgery Scissors, Integra™ Miltex®
15.2 cm (6"), 6 mm
Price
$1,697.17
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Straight with sharp points and round handles.
Specifications
  • Longueur:
    15.2 cm (6")
  • Blade Length:
    6 mm