Iris Dressing Forceps, OR Grade, Sklar
Supplier:   Sklar
Iris Dressing Forceps, OR Grade, Sklar
Supplier:  Sklar
Certificates
Iris Dressing Forceps, OR Grade, Sklar
Iris Dressing Forceps, OR Grade, Sklar
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar® Iris Dressing Forceps are thumb style forceps commonly used for placing or removing dressings during ophthalmic procedures.
Although Dressing Forceps were originally designed to specifically hold materials such as gauze, they have come to have a variety of additional functions
Censitrac Ready products are industry standard and constructed of stainless steel.
They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing.
 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Iris Dressing Forceps, OR Grade
Informations sur le produit
  • Désignation
    Grade
    Matière
    Longueur
    Tip Style
    Handle
    Curvature
    Sterile
    Availability
    Price
  • Iris Dressing Forceps
    Premium OR-Grade
    Stainless steel
    10.2 cm (4")
    Serrated
    Thumb
    Fully Curved
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.29
    $202.29
    Prix promotionnel pour les nouveaux clients
    Each
       $202.29
    Supplier Number: 66-3140
  • Iris Dressing Forceps
    Premium OR-Grade
    Stainless steel
    10.2 cm (4")
    Serrated
    Thumb
    Half Curved
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $185.81
    $185.81
    Prix promotionnel pour les nouveaux clients
    Each
       $185.81
    Supplier Number: 66-2940
  • Iris Dressing Forceps
    Premium OR-Grade
    Stainless steel
    10.2 cm (4")
    Serrated
    Thumb
    Straight
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $178.21
    $178.21
    Prix promotionnel pour les nouveaux clients
    Each
       $178.21
    Supplier Number: 66-2740
  • Iris Dressing Forceps
    Premium OR-Grade
    Stainless steel
    10.2 cm (4")
    Serrated
    Thumb
    Straight
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $363.45
    $363.45
    Prix promotionnel pour les nouveaux clients
    Each
       $363.45
    Supplier Number: 66-2840