Reverse-Action Tweezers, EXCELTA®
Supplier:   Excelta
Reverse-Action Tweezers, EXCELTA®
Supplier:  Excelta
Certificates
Reverse-Action Tweezers, EXCELTA®
Reverse-Action Tweezers, EXCELTA®
6 Options Available Below
Product Details & Documents
The reverse action tweezers are closed at rest, allowing for the operator to hold an item without any finger pressure on the tweezer.
  • Will hold parts without finger pressure for extended time
  • Tool determines pressure on component, not operator
Reverse action tweezers are normally closed at rest. This allows the item to be held without hand pressure for any length of time. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Product Family Options
Curved
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Length
    Tip Style
    Tip Width
    Curvature
    Availability
    Price
  • Tweezers, curved fine point, reverse action, 1 star quality
    Stainless steel, anti-magnetic
    112.5 mm (4.5")
    Curved Reverse Action Fine Points
    45° curved
    Discontinued
    Supplier Number: -
  • Tweezers, reverse action, curved, 3 star quality, wooden grips
    Stainless steel 316L, anti-magnetic
    150 mm (6")
    2 mm (0.08")
    Curved Strong Tip, Wood Grips
    0.08" (0.2 mm)
    45° curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $303.45
    $303.45
    Each
       $303.45
    Supplier Number: 30-SA
Oblique
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Length
    Tip Style
    Tip Width
    Curvature
    Availability
    Price
  • Tweezers, reverse action, oblique , 3 star quality
    Stainless steel 316L, anti-magnetic
    112.5 mm (4.5")
    0.2 mm (0.008")
    Oblique Fine Tip
    0.012" (0.3 mm)
    45° oblique
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.88
    $191.88
    Each
       $191.88
    Supplier Number: 31-SA-C
Straight
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Length
    Tip Style
    Tip Width
    Curvature
    Availability
    Price
  • Tweezers, fine point, reverse action, 1 star quality
    Stainless steel, anti-magnetic
    106.25 mm (4.25")
    Straight Reverse Action Fine Points
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.04
    $44.04
    Each
       $44.04
    Supplier Number: DN-3C-SA-SE
  • Tweezers, tapered flat point, reverse action, 1 star quality
    Stainless steel, anti-magnetic
    118.75 mm (4.75")
    Straight Duck-Bill Tip
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.94
    $35.94
    Each
       $35.94
    Supplier Number: DN-2A-SA-SE
  • Tweezers, reverse action, 3 star quality
    Stainless steel 316L, anti-magnetic
    125 mm (5")
    0.2 mm (0.008")
    Straight Fine Tip
    0.012" (0.3 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $182.88
    $182.88
    Each
       $182.88
    Supplier Number: 31-SA
  • Tweezers, reverse action, 3 star quality, wooden grips
    Stainless steel 316L, anti-magnetic
    150 mm (6")
    1.75 mm (0.070")
    Straight Strong Tip, Wooden Grips
    0.09" (2.25 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $232.50
    $232.50
    Each
       $232.50
    Supplier Number: 29-SA