O'Brien Stitch Scissors, Sklar
Catalogue # 89025-324
Fournisseur: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:O'Brien Stitch Scissors, Premium OR-Grade
- Length:9.5 cm (3.75")
- Material:Stainless steel
- Curvature:Angled
- Cat. no.:89025-324
- Supplier No.:22-2837
- Blade Type:Sharp/Sharp, Smooth
- Handle:Finger Rings
Spécifications
À propos de cet article
O'Brien Stitch Scissors are primarily used for suture removal. These scissors have sharp, fine, angled tips that are used to snip the suture rather than lifting the suture slightly first, as with Spencer or Littauer stitch scissors.
- Made of stainless steel
- Autoclavable
They are often used in dental procedures where lifting the suture may be unpractical due to location and space limitations.
Censitrac Ready products are industry standard.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.