VWR® Hemostatic Forceps
À propos de cet article
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
- Stainless steel fabrication
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
2 Options Available Below
Product Details & Documents
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
- Stainless steel fabrication
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Curved127 mm (5")Log in pour la disponibilitéPack of 10Pack of 10 - $2,151.60$2,151.60Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 50Case of 50 - $10,758.00$10,758.00Prix promotionnel pour les nouveaux clientsEach$215.16Pack of 10$2,151.60Case of 50$10,758.00
- Straight127 mm (5")Log in pour la disponibilitéPack of 10Pack of 10 - $614.16$614.16Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 50Case of 50 - $3,070.80$3,070.80Prix promotionnel pour les nouveaux clientsEach$61.42Pack of 10$614.16Case of 50$3,070.80