VWR® Specimen Forceps
Supplier: VWR International
Jump to:
About this item
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Stainless steel construction
- Straight tips
- Easy to use
Durable specimen forceps are built to withstand years of regular use.
2 Options Available Below
Product Details & Documents
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Stainless steel construction
- Straight tips
- Easy to use
Durable specimen forceps are built to withstand years of regular use.
Product Family Options
Product Information
- LongueurAvailabilityPrice
- 254 mm (10")Each$91.64Pack of 12$1,099.68Case of 60$5,498.40
- 305 mm (12")Each$100.05Pack of 12$1,200.58Case of 60$6,002.88