Axygen® Macrovolume Pipette Tips
10 ml, -, Clear, 100 Tips/Pack, 20 Pack/case
Catalog # 89005-641
Supplier: Corning
Fournisseur #: T-10ML-C
Jump to:
Product Details & Documents
These tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most brands of pipettors.
- These polypropylene tips are RNase-/DNase-free
- Nonpyrogenic
- Compatible with Gilson P10 ml and MLA pipettors
- Compatible with Eppendorf® pipettors
- Eppendorf® is a registered trademark of Eppendorf SE
They are available in Maxymum Recovery, beveled, non sterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
Spécifications
- Volume:10 ml
- Tip Color:Clear
- Packed:100 Tips/Pack, 20 Pack/case
- Sterile:-