Analytical Balances, Advanced MR
À propos de cet article
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
- Guided leveling
- Four interfaces: USB-A, USB-C, RS232, Ethernet
- Bluetooth option
User management within the 4.5" touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
3 Options Available Below
Product Details & Documents
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
- Guided leveling
- Four interfaces: USB-A, USB-C, RS232, Ethernet
- Bluetooth option
User management within the 4.5" touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
Specifications for Product Family
Spécifications
- Catalog No:
- 77323-128
- 77323-132
- 77323-136
- CA77323-128
- CA77323-132
- CA77323-136
- Modèle:
- MR104
- MR204
- MR304
- MR104
- MR204
- MR304
- Communication interface:
- USB-A, USB-C, RS232, Ethernet
- USB-A, USB-C, RS232, Ethernet
- USB-A, USB-C, RS232, Ethernet
- USB-A, USB-C, RS232, Ethernet
- USB-A, USB-C, RS232, Ethernet
- USB-A, USB-C, RS232, Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Spécifications
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- MR104120 g0.2 mgØ 90 mm0.1 mg2 s0.1 mgDiscontinued-Supplier Number: 30666203Spécifications:
More
Specifications
Cat. No.77323-128ModèleMR104Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz - MR204220 g0.2 mgØ 90 mm0.1 mg2 s0.1 mgDiscontinued-Supplier Number: 30666206Spécifications:
More
Specifications
Cat. No.77323-132ModèleMR204Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz - MR304320 g0.2 mgØ 90 mm0.1 mg2 s0.1 mgDiscontinued-Supplier Number: 30666209Spécifications:
More
Specifications
Cat. No.77323-136ModèleMR304Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666203Spécifications:
More
Specifications
Cat. No.CA77323-128ModèleMR104Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666206Spécifications:
More
Specifications
Cat. No.CA77323-132ModèleMR204Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666209Spécifications:
More
Specifications
Cat. No.CA77323-136ModèleMR304Communication interfaceUSB-A, USB-C, RS232, EthernetVoltage100 - 240 V, 50/60 Hz