Standard LA Precision Balances
About this item
METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.
- Front level indicator
- Overload protection
- Metal base, ABS top housing
- RS232 interface
- Bluetooth option
- Transfer weight function for easy data transfer
Increase efficiency in daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.
4 Options Available Below
Product Details & Documents
METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.
- Front level indicator
- Overload protection
- Metal base, ABS top housing
- RS232 interface
- Bluetooth option
- Transfer weight function for easy data transfer
Increase efficiency in daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.
Specifications for Product Family
Specifications
- Catalog No:
- CA77323-286
- 77323-286
- CA77323-282
- 77323-282
- CA77323-288
- 77323-288
- CA77323-284
- 77323-284
- Modèle:
- LA203E
- LA203E
- LA403E
- LA403E
- LA2002E
- LA2002E
- LA4002E
- LA4002E
- Communication interface:
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Specifications
Product Family Options
Product Information
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- Supplier Number: 30853154Specifications:
More
Specifications
Cat. No.CA77323-286ModèleLA203ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - LA203E220 g2 mgØ 100 mm1 mg3 s1 mgDiscontinued-Supplier Number: 30853154Specifications:
More
Specifications
Cat. No.77323-286ModèleLA203ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - Supplier Number: 30823722Specifications:
More
Specifications
Cat. No.CA77323-282ModèleLA403ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - LA403E420 g2 mgØ 100 mm1 mg3 s1 mgDiscontinued-Supplier Number: 30823722Specifications:
More
Specifications
Cat. No.77323-282ModèleLA403ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - Supplier Number: 30853155Specifications:
More
Specifications
Cat. No.CA77323-288ModèleLA2002ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - LA2002E2200 g20 mgØ 180 mm10 mg3 s10 mgDiscontinued-Supplier Number: 30853155Specifications:
More
Specifications
Cat. No.77323-288ModèleLA2002ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - Supplier Number: 30823723Specifications:
More
Specifications
Cat. No.CA77323-284ModèleLA4002ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz - LA4002E4200 g20 mgØ 180 mm10 mg3 s10 mgDiscontinued-Supplier Number: 30823723Specifications:
More
Specifications
Cat. No.77323-284ModèleLA4002ECommunication interfaceUSB-A, RS232Voltage100 - 240 V, 50/60 Hz