Advanced MR Precision Balances
À propos de cet article
METTLER TOLEDO Advanced MR Precision Balances feature a precise weigh cell and FACT automatic internal adjustment for highly accurate results.
- User management
- Guided leveling
- Statistical data analysis
- 4 interfaces: USB-A, USB-C, RS232, Ethernet
- Bluetooth option
- Full metal housing
- Overload protection
The 4.5" touchscreen with intuitive user management provides easy access to 9 built-in applications and increases efficiency in the workflow. Guided leveling, multiple connectivity options and easy to clean balance features reduces time spent on routine tasks. Designed with IP 43 protection and full metal housing to ensure long lasting performance. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
10 Options Available Below
Product Details & Documents
METTLER TOLEDO Advanced MR Precision Balances feature a precise weigh cell and FACT automatic internal adjustment for highly accurate results.
- User management
- Guided leveling
- Statistical data analysis
- 4 interfaces: USB-A, USB-C, RS232, Ethernet
- Bluetooth option
- Full metal housing
- Overload protection
The 4.5" touchscreen with intuitive user management provides easy access to 9 built-in applications and increases efficiency in the workflow. Guided leveling, multiple connectivity options and easy to clean balance features reduces time spent on routine tasks. Designed with IP 43 protection and full metal housing to ensure long lasting performance. When balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
Specifications for Product Family
Spécifications
- Catalog No:
- CA77323-140
- 77323-140
- CA77323-144
- 77323-144
- CA77323-148
- 77323-148
- CA77323-152
- 77323-152
- CA77323-156
- 77323-156
- 77323-160
- CA77323-160
- 77323-164
- CA77323-164
- 77323-168
- CA77323-168
- 77323-172
- CA77323-172
- 77323-176
- CA77323-176
- Modèle:
- MR203
- MR203
- MR303
- MR303
- MR503
- MR503
- MR603
- MR603
- MR1002
- MR1002
- MR2002
- MR2002
- MR3002
- MR3002
- MR4002
- MR4002
- MR6002
- MR6002
- MR6001
- MR6001
- Communication interface:
- USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet - USB-A, USB-C, RS232,
Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Spécifications
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- Supplier Number: 30666212Spécifications:
More
Specifications
Cat. No.CA77323-140ModèleMR203Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR203220 g2 mgØ 120 mm1 mg1.5 s1 mgDiscontinued-Supplier Number: 30666212Spécifications:
More
Specifications
Cat. No.77323-140ModèleMR203Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666215Spécifications:
More
Specifications
Cat. No.CA77323-144ModèleMR303Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR303320 g2 mgØ 120 mm1 mg1.5 s1 mgDiscontinued-Supplier Number: 30666215Spécifications:
More
Specifications
Cat. No.77323-144ModèleMR303Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666218Spécifications:
More
Specifications
Cat. No.CA77323-148ModèleMR503Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR503520 g2 mgØ 120 mm1 mg1.5 s1 mgDiscontinued-Supplier Number: 30666218Spécifications:
More
Specifications
Cat. No.77323-148ModèleMR503Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666221Spécifications:
More
Specifications
Cat. No.CA77323-152ModèleMR603Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR603620 g2 mgØ 120 mm1 mg1.5 s1 mgDiscontinued-Supplier Number: 30666221Spécifications:
More
Specifications
Cat. No.77323-152ModèleMR603Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666224Spécifications:
More
Specifications
Cat. No.CA77323-156ModèleMR1002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR10021.2 kg20 mg180×180 mm10 mg1 s10 mgDiscontinued-Supplier Number: 30666224Spécifications:
More
Specifications
Cat. No.77323-156ModèleMR1002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR20022.2 kg20 mg180×180 mm10 mg1 s10 mgDiscontinued-Supplier Number: 30666227Spécifications:
More
Specifications
Cat. No.77323-160ModèleMR2002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666227Spécifications:
More
Specifications
Cat. No.CA77323-160ModèleMR2002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR30023.2 kg20 mg180×180 mm10 mg1 s10 mgDiscontinued-Supplier Number: 30666230Spécifications:
More
Specifications
Cat. No.77323-164ModèleMR3002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666230Spécifications:
More
Specifications
Cat. No.CA77323-164ModèleMR3002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR40024.2 kg20 mg180×180 mm10 mg1 s10 mgDiscontinued-Supplier Number: 30666233Spécifications:
More
Specifications
Cat. No.77323-168ModèleMR4002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666233Spécifications:
More
Specifications
Cat. No.CA77323-168ModèleMR4002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR60026.2 kg20 mg180×180 mm10 mg1 s10 mgDiscontinued-Supplier Number: 30666236Spécifications:
More
Specifications
Cat. No.77323-172ModèleMR6002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666236Spécifications:
More
Specifications
Cat. No.CA77323-172ModèleMR6002Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - MR60016.2 kg60 mg180×180 mm0.1 g1 s80 mgDiscontinued-Supplier Number: 30666239Spécifications:
More
Specifications
Cat. No.77323-176ModèleMR6001Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30666239Spécifications:
More
Specifications
Cat. No.CA77323-176ModèleMR6001Communication interfaceUSB-A, USB-C, RS232,
EthernetVoltage100 - 240 V, 50/60 Hz