Advanced MX Precision Balances
À propos de cet article
METTLER TOLEDO Advanced MX Precision Balances feature a large weighing pan, robust design, and precise weigh cell with FACT automatic internal adjustment.
- SmartPan
- Statistical data analysis
- User management; Leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
- Full metal housing with overload protection
The 7" intuitive touchscreen with user management provides automatic data logging, statistical analysis and easy access to 9 built-in weighing applications. Transfer data to a printer or computer for error-free documentation. Built with IP 54 protection and robust construction, these balances will deliver high quality performance for years to come. Enable the power-saving mode when balance is not in use to saves costs and reduces energy consumption.
4 Options Available Below
Product Details & Documents
METTLER TOLEDO Advanced MX Precision Balances feature a large weighing pan, robust design, and precise weigh cell with FACT automatic internal adjustment.
- SmartPan
- Statistical data analysis
- User management; Leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
- Full metal housing with overload protection
The 7" intuitive touchscreen with user management provides automatic data logging, statistical analysis and easy access to 9 built-in weighing applications. Transfer data to a printer or computer for error-free documentation. Built with IP 54 protection and robust construction, these balances will deliver high quality performance for years to come. Enable the power-saving mode when balance is not in use to saves costs and reduces energy consumption.
Specifications for Product Family
Spécifications
- Catalog No:
- 77323-112
- CA77323-112
- 77323-116
- CA77323-116
- CA77323-120
- 77323-120
- CA77323-124
- 77323-124
- Modèle:
- MX12001L
- MX12001L
- MX16001L
- MX16001L
- MX32001L
- MX32001L
- MX32000L
- MX32000L
- Communication interface:
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Spécifications
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- MX12001L12.2 kg200 mg352×246 mm0.1 g1.5 s80 mgDiscontinued-Supplier Number: 30665188Spécifications:
More
Specifications
Cat. No.77323-112ModèleMX12001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665188Spécifications:
More
Specifications
Cat. No.CA77323-112ModèleMX12001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX16001L16.2 kg200 mg352×246 mm0.1 g1.5 s80 mgDiscontinued-Supplier Number: 30665191Spécifications:
More
Specifications
Cat. No.77323-116ModèleMX16001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665191Spécifications:
More
Specifications
Cat. No.CA77323-116ModèleMX16001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665194Spécifications:
More
Specifications
Cat. No.CA77323-120ModèleMX32001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX32001L32.2 kg250 mg352×246 mm0.1 g1.5 s80 mgDiscontinued-Supplier Number: 30665194Spécifications:
More
Specifications
Cat. No.77323-120ModèleMX32001LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665197Spécifications:
More
Specifications
Cat. No.CA77323-124ModèleMX32000LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX32000L32.2 kg300 mg352×246 mm1 g1.2 s600 mgDiscontinued-Supplier Number: 30665197Spécifications:
More
Specifications
Cat. No.77323-124ModèleMX32000LCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz