Advanced MX Precision Balances
À propos de cet article
METTLER TOLEDO Advanced MX Precision Balances feature a precise weigh cell and automatic internal adjustment. The SmartPan stabilizes in any weighing environment, ensuring consistently accurate results.
- FACT automatic internal adjustment
- Statistical data analysis
- User management; leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
- Full metal housing with overload protection
Exceptional user management within the 7" touchscreen allows easy access to 9 built-in weighing applications, assigns user access rights and provides automatic data logging. These balances are designed with full metal housing and IP 54 protection to defend against environmental influences and rough handling. The power-saving mode can be enabled when the balance is not in use to save costs and reduce energy usage.
13 Options Available Below
Product Details & Documents
METTLER TOLEDO Advanced MX Precision Balances feature a precise weigh cell and automatic internal adjustment. The SmartPan stabilizes in any weighing environment, ensuring consistently accurate results.
- FACT automatic internal adjustment
- Statistical data analysis
- User management; leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
- Full metal housing with overload protection
Exceptional user management within the 7" touchscreen allows easy access to 9 built-in weighing applications, assigns user access rights and provides automatic data logging. These balances are designed with full metal housing and IP 54 protection to defend against environmental influences and rough handling. The power-saving mode can be enabled when the balance is not in use to save costs and reduce energy usage.
Specifications for Product Family
Spécifications
- Catalog No:
- CA77323-060
- 77323-060
- CA77323-064
- 77323-064
- CA77323-068
- 77323-068
- CA77323-072
- 77323-072
- CA77323-076
- 77323-076
- CA77323-080
- 77323-080
- CA77323-096
- 77323-096
- CA77323-084
- 77323-084
- CA77323-088
- 77323-088
- CA77323-092
- 77323-092
- 77323-104
- CA77323-104
- 77323-108
- CA77323-108
- 77323-100
- CA77323-100
- Modèle:
- MX303
- MX303
- MX303N
- MX303N
- MX603
- MX603
- MX603N
- MX603N
- MX1203
- MX1203
- MX1203N
- MX1203N
- MX6002DR
- MX6002DR
- MX2002
- MX2002
- MX4002
- MX4002
- MX6002
- MX6002
- MX6001
- MX6001
- MX8001
- MX8001
- MX12002
- MX12002
- Communication interface:
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
Spécifications
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- Supplier Number: 30665149Spécifications:
More
Specifications
Cat. No.CA77323-060ModèleMX303Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX303320 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665149Spécifications:
More
Specifications
Cat. No.77323-060ModèleMX303Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665152Spécifications:
More
Specifications
Cat. No.CA77323-064ModèleMX303NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX303N320 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665152Spécifications:
More
Specifications
Cat. No.77323-064ModèleMX303NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665155Spécifications:
More
Specifications
Cat. No.CA77323-068ModèleMX603Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX603620 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665155Spécifications:
More
Specifications
Cat. No.77323-068ModèleMX603Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665158Spécifications:
More
Specifications
Cat. No.CA77323-072ModèleMX603NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX603N620 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665158Spécifications:
More
Specifications
Cat. No.77323-072ModèleMX603NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665161Spécifications:
More
Specifications
Cat. No.CA77323-076ModèleMX1203Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX12031220 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665161Spécifications:
More
Specifications
Cat. No.77323-076ModèleMX1203Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665164Spécifications:
More
Specifications
Cat. No.CA77323-080ModèleMX1203NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX1203N1220 g2 mg127×127 mm1 mg1.5 s0.9 mgDiscontinued-Supplier Number: 30665164Spécifications:
More
Specifications
Cat. No.77323-080ModèleMX1203NCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665176Spécifications:
More
Specifications
Cat. No.CA77323-096ModèleMX6002DRCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX6002DR1.2/6.2 kg30 mg172×205 mm0.01/0.1 g1.2 s8 mgDiscontinued-Supplier Number: 30665176Spécifications:
More
Specifications
Cat. No.77323-096ModèleMX6002DRCommunication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665167Spécifications:
More
Specifications
Cat. No.CA77323-084ModèleMX2002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX20022.2 kg20 mg172×205 mm0.01 g1.2 s8 mgDiscontinued-Supplier Number: 30665167Spécifications:
More
Specifications
Cat. No.77323-084ModèleMX2002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665170Spécifications:
More
Specifications
Cat. No.CA77323-088ModèleMX4002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX40024.2 kg20 mg172×205 mm0.01 g1.2 s8 mgDiscontinued-Supplier Number: 30665170Spécifications:
More
Specifications
Cat. No.77323-088ModèleMX4002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665173Spécifications:
More
Specifications
Cat. No.CA77323-092ModèleMX6002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX60026.2 kg20 mg172×205 mm0.01 g1.2 s8 mgDiscontinued-Supplier Number: 30665173Spécifications:
More
Specifications
Cat. No.77323-092ModèleMX6002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX60016.2 kg60 mg172×205 mm0.1 g1 s50 mgDiscontinued-Supplier Number: 30665182Spécifications:
More
Specifications
Cat. No.77323-104ModèleMX6001Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665182Spécifications:
More
Specifications
Cat. No.CA77323-104ModèleMX6001Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX80018.2 kg100 mg172×205 mm0.1 g1 s50 mgDiscontinued-Supplier Number: 30665185Spécifications:
More
Specifications
Cat. No.77323-108ModèleMX8001Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665185Spécifications:
More
Specifications
Cat. No.CA77323-108ModèleMX8001Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - MX1200212.2 kg20 mg172×205 mm0.01 g1.2 s8 mgDiscontinued-Supplier Number: 30665179Spécifications:
More
Specifications
Cat. No.77323-100ModèleMX12002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz - Supplier Number: 30665179Spécifications:
More
Specifications
Cat. No.CA77323-100ModèleMX12002Communication interfaceUSB-A, USB-C, EthernetVoltage100 - 240 V, 50/60 Hz