Sullivan Scissors, Sklar
À propos de cet article
Sklar's® Sullivan (Brophy) surgical scissors are designed to cut through the tissue of the gum
Censitrac Ready OR Grade Sullivan Scissors are industry standard and constructed of stainless steel.
Although designed for oral surgery.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
2 Options Available Below
Product Details & Documents
Sklar's® Sullivan (Brophy) surgical scissors are designed to cut through the tissue of the gum
Censitrac Ready OR Grade Sullivan Scissors are industry standard and constructed of stainless steel.
Although designed for oral surgery.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Sullivan Scissors
Informations sur le produit
- DésignationLongueurMatièreBlade TypeCurvatureHandleSterilityAvailabilityPrice
- Brophy (Sullivan) Scissors, Premium OR-Grade14.6 cm (5.75")Stainless steelSerratedCurvedFinger RingsNon sterileLog in pour la disponibilitéEach$406.84Supplier Number: 22-2755
- Brophy (Sullivan) Scissors, Premium OR-Grade15.2 cm (6")Stainless steelSharp/Sharp, SerratedStraightFinger RingsNon sterileLog in pour la disponibilitéEach$406.84Supplier Number: 22-2855