ContactUsLinkComponent
VWR® Dissecting Scissors, Curved Tip, 5"
Catalogue # 82027-586
Fournisseur:  VWR International
undefined
VWR® Dissecting Scissors, Curved Tip, 5"
Catalogue # 82027-586
Fournisseur:  VWR International
Spécifications
  • Length:
    127 mm (5")
  • Cat. no.:
    82027-586
Spécifications
À propos de cet article
VWR® Dissecting Scissors are stainless steel with fine, bent tips.
  • Stainless steel fabrication
  • Curved tips
Dissecting scissors are used for a variety of applications and are available in different lengths.