VWR® Dissecting Scissors, Sharp Tip, 4¹/₂" | Avantor
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
114 mm (4½")
Catalog # 82027-578
Supplier:   VWR International
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
Catalog # 82027-578
Supplier:  VWR International
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
114 mm (4½")
Price
$47.23
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each Pack of 10 Case of 50
Product Details & Documents
VWR® Dissecting Scissors are stainless steel with fine, straight tips.
  • Stainless steel fabrication
  • Fine, straight, sharp tips
Dissecting scissors are used for a variety of applications and are available in different lengths.
Spécifications
  • Longueur:
    114 mm (4½")