VITLAB® HDPE Buckets, BrandTech
Fournisseur: Brandtech
Certificats
À propos de cet article
Buckets feature a hinged handle and graduation markings.
- HDPE buckets with handles
- Snap-on PP lids sold separately
- Durable
These VITLAB® buckets are made of heavy-duty high-density polyethylene for durability and chemical resistance. A hinged handle provides easy gripping and stability. Graduations provide easy estimation of content volumes.
Frequently Bought Together