Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Plastic Forceps
Catalog # 30049-122
Supplier: Thermo Fisher Scientific
Fournisseur #: 6320-0010
Nalgene® Polypropylene Locking Forceps, Thermo Scientific
Catalog # 30049-122
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
All-plastic forceps with double-action hinges for extra leverage and long life
Scissors-type handles with ratchet for locking tips shut. Interlocking teeth for tight grip. Provide a useful clamp for chromatography columns. Durable blue polypropylene is autoclavable for repeated use. Ideal for economical, disposable applications.
Spécifications
- Désignation:Plastic Forceps