Trypsinizing Flasks, Bellco Glass
Trypsinizing Flasks, Bellco Glass
Supplier:   Bellco Glass
Trypsinizing Flasks, Bellco Glass
Supplier:  Bellco Glass
Certificates
Trypsinizing Flasks, Bellco Glass
2 Options Available Below
Product Details & Documents
Manufactured from Type 1 Class A Borosilicate Glass.
  • Designed with deep side baffles for optimal agitation
  • Large bottom surface for easy access with Bellco large orifice pipette
Product Family Options
Product Information
  • Désignation
    O.D.×H
    Availability
    Price
  • Trypsinizing flask, 50 ml
    50×80 mm
    Log in for availability
    Log in for availability
    Case of 3 (50ml)
    Case of 3 (50ml) - $5,049.00
    $5,049.00
    Case of 3 (50ml)
       $5,049.00
    Minimum order quantity is 1
    Supplier Number: 1992-00050
    Specifications:
    Marketsource
    Marketsource
  • Trypsinizing flask, 250 ml
    80×140 mm
    Log in for availability
    Log in for availability
    Case of 3 (250ml)
    Case of 3 (250ml) - $6,703.10
    $6,703.10
    Case of 3 (250ml)
       $6,703.10
    Supplier Number: 1992-00250