Stirring Bars, Bellco
Marketsource
Supplier:   Bellco Glass
Stirring Bars, Bellco
Supplier:  Bellco Glass
Marketsource
Certificates
Stirring Bars, Bellco
5 Options Available Below
Product Details & Documents
Molded, seamless PTFE construction is ideal for procedures using all Bellco magnetic stirrers. Excellent for mixing media and buffer solutions.
Product Family Options
Product Information
  • Longueur
    Ø
    Color
    Availability
    Price
  • 25.4 mm
    7.6 mm
    White
    Log in for availability
    Log in for availability
    Case of 6 (1/Itm/Bx)
    Case of 6 (1/Itm/Bx) - $5,389.17
    $5,389.17
    Case of 6 (1/Itm/Bx)
       $5,389.17
    Supplier Number: 1975-00100
    Specifications:
    Marketsource
    Marketsource
  • 41.3 mm
    9.5 mm
    White
    Log in for availability
    Log in for availability
    Case of 6 (1/Itm/Bx)
    Case of 6 (1/Itm/Bx) - $5,518.51
    $5,518.51
    Case of 6 (1/Itm/Bx)
       $5,518.51
    Supplier Number: 1975-00158
  • 50.8 mm
    9.5 mm
    White
    Log in for availability
    Log in for availability
    Case of 6 (1/Itm/Bx)
    Case of 6 (1/Itm/Bx) - $5,869.64
    $5,869.64
    Case of 6 (1/Itm/Bx)
       $5,869.64
    Supplier Number: 1975-00200
    Specifications:
    Marketsource
    Marketsource
  • 76.2 mm
    12.1 mm
    White
    Log in for availability
    Log in for availability
    Case of 3 (1/Itm/Bx)
    Case of 3 (1/Itm/Bx) - $6,080.79
    $6,080.79
    Case of 3 (1/Itm/Bx)
       $6,080.79
    Supplier Number: 1975-00300
  • 82.6 mm
    19 mm
    White
    Log in for availability
    Log in for availability
    Case of 2 (1/Itm/Bx)
    Case of 2 (1/Itm/Bx) - $5,013.17
    $5,013.17
    Case of 2 (1/Itm/Bx)
       $5,013.17
    Supplier Number: 1975-00314
    Specifications:
    Marketsource
    Marketsource