VWR® Micro-Adson Forceps
127 mm (5")
Supplier: VWR International
Jump to:
Product Details & Documents
Micro-Adson forceps are for holding and manipulating delicate tissues.
- Stainless steel construction
- Micro-Adson type
- Serrated jaws
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are extremely narrow.
Spécifications
- Longueur:127 mm (5")