VWR® Smooth Jaw Forceps
203 mm (8")
Catalog # 82027-448
Supplier: VWR International
Jump to:
Product Details & Documents
Stainless steel with straight, smooth jaws are ideal for use when you need an extra pair of hands on any number of craft applications.
- Stainless steel construction
- Straight, smooth jaws
- Adjustable tension
- Quick release
Smooth jaws will not make any mark on fabric.
Spécifications
- Longueur:203 mm (8")